missing translation for 'onlineSavingsMsg'
Learn More

o-Orsellinic acid hydrate, 98%

Catalog No. p-7077414
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453290010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC453290010 Supplier Thermo Scientific Chemicals Supplier No. 453290010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 480-64-8 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 480-64-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD16661187
InChI Key AMKYESDOVDKZKV-UHFFFAOYSA-N
Synonym orsellinic acid, orsellic acid, o-orsellinic acid, orcinolcarboxylic acid, 4,6-dihydroxy-o-toluic acid, unii-11xla0494b, benzoic acid, 2,4-dihydroxy-6-methyl, 2,4-dihydroxy-6-methyl-benzoic acid, orsellinate, 4,6-dihydroxy-2-methylbenzoic acid
PubChem CID 68072
ChEBI CHEBI:32807
IUPAC Name 2,4-dihydroxy-6-methylbenzoic acid
SMILES CC1=CC(O)=CC(O)=C1C(O)=O

Specifications

Melting Point 152.0°C to 153.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 186.17
Percent Purity 98%
Chemical Name or Material o-Orsellinic acid monohydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only