Learn More
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 97%, Thermo Scientific™
$131.47 - $414.47
Chemical Identifiers
| CAS | 105832-38-0 |
|---|---|
| Molecular Formula | C9H16N3O3·BF4 |
| Molecular Weight (g/mol) | 301.05 |
| InChI Key | YEBLHMRPZHNTEK-UHFFFAOYSA-N |
| Synonym | tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781 |
| PubChem CID | 9857522 |
| IUPAC Name | [dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;tetrafluoroborate |
| SMILES | [B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC303190010
|
N/A | 1g | Glass bottle |
Each for $131.47
|
|
||||
|
AC303190050
|
N/A | 5g | Glass bottle |
Each for $414.47
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Coupling reagentChemical Identifiers
| 105832-38-0 | |
| 301.05 | |
| tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781 | |
| [dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;tetrafluoroborate |
| C9H16N3O3·BF4 | |
| YEBLHMRPZHNTEK-UHFFFAOYSA-N | |
| 9857522 | |
| [B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O |
Specifications
| 105832-38-0 | |
| 100.0 | |
| Authentic | |
| C9H16N3O3·BF4 | |
| YEBLHMRPZHNTEK-UHFFFAOYSA-N | |
| [dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;tetrafluoroborate | |
| 9857522 | |
| 97% |
| 96.0 | |
| 34.47 to 37.34% C, 13.40 to 14.52% N | |
| 97% | |
| tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781 | |
| [B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O | |
| 301.05 | |
| 301.05 | |
| O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON
GHS Signal Word: Warning
RUO – Research Use Only