missing translation for 'onlineSavingsMsg'
Learn More

O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 97%

Catalog No. AC303190010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303190010 1 g Glass bottle
AC303190050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC303190010 Supplier Thermo Scientific Chemicals Supplier No. 303190010

CAS: 105832-38-0 | C9H16BF4N3O3 | 301.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 105832-38-0
Molecular Formula C9H16BF4N3O3
Molecular Weight (g/mol) 301.05
MDL Number MFCD00077875
InChI Key YEBLHMRPZHNTEK-UHFFFAOYSA-N
Synonym tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781
PubChem CID 9857522
IUPAC Name [dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 194.0°C to 200.0°C
Color White
Elemental Analysis 34.47 to 37.34% C, 13.40 to 14.52% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 301.05
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
missing translation for 'healthHazard2' GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
missing translation for 'healthHazard3' GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON

RUO – Research Use Only