missing translation for 'onlineSavingsMsg'
Learn More

O,N,N'-Triisopropylisourea 98.0+%, TCI America™

Catalog No. T312325ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T312325ML 25 mL
T31235ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T312325ML Supplier TCI America Supplier No. T312325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63460-32-2
Molecular Formula C10H22N2O
Molecular Weight (g/mol) 186.299
InChI Key RHNDDRWPYPWKNW-UHFFFAOYSA-N
PubChem CID 11332901
IUPAC Name propan-2-yl N,N'-di(propan-2-yl)carbamimidate
SMILES CC(C)NC(=NC(C)C)OC(C)C

Specifications

Color Colorless
Boiling Point 67°C
Quantity 25 mL
Synonym Isopropyl N,N′-Diisopropylcarbamimidate, N,N′-Diisopropylcarbamimidic Acid Isopropyl Ester
Formula Weight 186.30
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material O,N,N′-Triisopropylisourea
TSCA No
Recommended Storage Refrigerator