accessibility menu, dialog, popup

UserName

CAS RN 63460-32-2

IUPAC Name: N,N'-bis(propan-2-yl)(propan-2-yloxy)methanimidamide
Synonyms: N,N'-diisopropylisopropoxymethanimidamide
Molecular Weight (g/mol): 186.3
Molecular Formula: C10H22N2O
InChi Key: RHNDDRWPYPWKNW-UHFFFAOYSA-N
SMILES: CC(C)NC(OC(C)C)=NC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results