missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Methylisourea Sulfate 98.0+%, TCI America™
Supplier: TCI America M0411100G
Specifications
| O-Methylisourea Sulfate | |
| 167°C | |
| C4H14N4O6S | |
| 100 g | |
| QSCPQKVWSNUJLJ-UHFFFAOYSA-N | |
| methyl carbamimidate;sulfuric acid | |
| 11357018 | |
| ≥98.0% (N) |
| 52328-05-9 | |
| White | |
| MFCD00040594 | |
| o-methylisourea hemisulfate, 2-methylisouronium sulfate, o-methylisourea hemisulfate salt, bis methoxymethanimidamide ; sulfuric acid, carbamimidic acid, methyl ester, sulfate 2:1, omi™, o-methylpseudourea sulfate, 0-methylisourea hemisulfate, o-methylisourea hemisulphate, bis 2-methylisouronium sulphate | |
| COC(=N)N.COC(=N)N.OS(=O)(=O)O | |
| 246.238 | |
| 246.24 | |
| Crystalline Powder |
Chemical Identifiers
| 52328-05-9 | |
| 246.238 | |
| QSCPQKVWSNUJLJ-UHFFFAOYSA-N | |
| 11357018 | |
| COC(=N)N.COC(=N)N.OS(=O)(=O)O |
| C4H14N4O6S | |
| MFCD00040594 | |
| o-methylisourea hemisulfate, 2-methylisouronium sulfate, o-methylisourea hemisulfate salt, bis methoxymethanimidamide ; sulfuric acid, carbamimidic acid, methyl ester, sulfate 2:1, omi™, o-methylpseudourea sulfate, 0-methylisourea hemisulfate, o-methylisourea hemisulphate, bis 2-methylisouronium sulphate | |
| methyl carbamimidate;sulfuric acid |
Safety and Handling
TSCA : No