missing translation for 'onlineSavingsMsg'
Learn More

O-Methylisourea Sulfate 98.0+%, TCI America™

Catalog No. p-7132151
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M041110G 10 g
M0411100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M041110G Supplier TCI America Supplier No. M041110G
Only null left

Chemical Identifiers

CAS 52328-05-9
Molecular Formula C4H14N4O6S
Molecular Weight (g/mol) 246.238
MDL Number MFCD00040594
InChI Key QSCPQKVWSNUJLJ-UHFFFAOYSA-N
Synonym o-methylisourea hemisulfate, 2-methylisouronium sulfate, o-methylisourea hemisulfate salt, bis methoxymethanimidamide ; sulfuric acid, carbamimidic acid, methyl ester, sulfate 2:1, omi™, o-methylpseudourea sulfate, 0-methylisourea hemisulfate, o-methylisourea hemisulphate, bis 2-methylisouronium sulphate
PubChem CID 11357018
IUPAC Name methyl carbamimidate;sulfuric acid
SMILES COC(=N)N.COC(=N)N.OS(=O)(=O)O

Specifications

Melting Point 167°C
Color White
Quantity 10 g
Formula Weight 246.24
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material O-Methylisourea Sulfate