missing translation for 'onlineSavingsMsg'
Learn More

O-Methylisourea hydrochloride, 98%

Catalog No. AAA1477330
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1477330 250 g
AAA1477309 10 g
AAA1477318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1477330 Supplier Thermo Scientific Chemicals Supplier No. A1477330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5329-33-9 | C2H7ClN2O | 110.541 g/mol

O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.

Solubility
Soluble in organic solvents, water, methanol and ethanol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 5329-33-9
Molecular Formula C2H7ClN2O
Molecular Weight (g/mol) 110.541
MDL Number MFCD00035043
InChI Key MUDVUWOLBJRUGF-UHFFFAOYSA-N
Synonym o-methylisourea hydrochloride, methyl carbamimidate hydrochloride, o-methyluronium chloride, 2-methylpseudourea hydrochloride, o-methyl-isourea hydrochloride, methoxymethanimidamide hydrochloride, carbamimidic acid, methyl ester, monohydrochloride, carbamimidic acid, methyl ester, hydrochloride 1:1, 2-methylpseudourea hcl, acmc-20ak09
PubChem CID 3083899
IUPAC Name methyl carbamimidate;hydrochloride
SMILES COC(=N)N.Cl

Specifications

Melting Point ∼116°C (decomposition)
Quantity 250 g
Beilstein 3909619
Solubility Information Soluble in organic solvents,water,methanol and ethanol.
Formula Weight 110.54
Percent Purity 98%
Chemical Name or Material O-Methylisourea hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only