CAS RN 5329-33-9
IUPAC Name:
hydrogen methoxymethanimidamide chloride
Synonyms:
hydrogen methoxymethanimidamide chloride
Molecular Weight (g/mol):
110.54
Molecular Formula:
C2H7ClN2O
InChi Key:
MUDVUWOLBJRUGF-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].COC(N)=N
1
–
1
of
1
results
1
Promotions Available
O-Methylisourea hydrochloride, 98%
O-Methylisourea hydrochloride, 98%
CAS: 5329-33-9 Molecular Formula: C2H7ClN2O Molecular Weight (g/mol): 110.541 MDL Number: MFCD00035043 InChI Key: MUDVUWOLBJRUGF-UHFFFAOYSA-N Synonym: o-methylisourea hydrochloride,methyl carbamimidate hydrochloride,o-methyluronium chloride,2-methylpseudourea hydrochloride,o-methyl-isourea hydrochloride,methoxymethanimidamide hydrochloride,carbamimidic acid, methyl ester, monohydrochloride,carbamimidic acid, methyl ester, hydrochloride 1:1,2-methylpseudourea hcl,acmc-20ak09 PubChem CID: 3083899 IUPAC Name: methyl carbamimidate;hydrochloride SMILES: COC(=N)N.Cl