missing translation for 'onlineSavingsMsg'
Learn More

O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. E13061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E13061G 1 g
E13065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E13061G Supplier TCI America Supplier No. E13061G

Chemical Identifiers

CAS 333717-40-1
Molecular Formula C10H17F6N4O3P
Molecular Weight (g/mol) 386.235
MDL Number MFCD01862832
InChI Key RKTBAMPZUATMIO-JCTPKUEWSA-N
Synonym HOTU
PubChem CID 57416861
IUPAC Name [[(Z)-(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CCOC(=O)C(=NOC(=[N+](C)C)N(C)C)C#N.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 132°C
Color White
Quantity 1 g
Formula Weight 386.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N′,N′-tetramethyluronium Hexafluorophosphate
TSCA No
Recommended Storage Refrigerator