missing translation for 'onlineSavingsMsg'
Learn More

o-Dianisidine dihydrochloride, For Enzymic, Spectrophotometric Determination, MilliporeSigma™ Supelco™

Catalog No. 111012178
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012178 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012178 Supplier Merck Emd Millipore Supplier No. D26791VL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Vial
Quantity 5 mg
Synonym o-Dianisidine reagent
Grade For enzymic, spectrophotometric determination