missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals o-Cresolphthalein Complexone, pure

Catalog No. AC182440050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC182440050 5 g Glass bottle
AC182440250 25 g Glass bottle
AC182441000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC182440050 Supplier Thermo Scientific Chemicals Supplier No. 182440050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2411-89-4 | C32H32N2O12 | 636.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 2411-89-4
Molecular Formula C32H32N2O12
Molecular Weight (g/mol) 636.61
MDL Number MFCD00005911
InChI Key IYZPEGVSBUNMBE-UHFFFAOYSA-N
Synonym o-Cresolphthalexon, Phthalein Complexon
PubChem CID 75485
IUPAC Name 2-[({5-[1-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
SMILES CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1

Specifications

Melting Point 181.0°C
Merck Index 15, 2566
Solubility Information Solubility in water: slightly soluble. Other solubilities: readily soluble in aqueous ammonia, readily soluble in organic solvents
Infrared Spectrum Authentic
Lambda Max 573 to 579nm (in 0.1N NaOH)
Formula Weight 636.6
Loss on Drying 10% max. (1 g, 105°C)
Grade Pure
Packaging Glass bottle
Suitability As indicator for complexometry: Passes Test
UV Spectrometry Complies
Color Brown to White
Quantity 5 g
Physical Form Powder
Chemical Name or Material o-Cresolphthalein Complexone, Pure
Show More Show Less
EINECSNumber 219-318-8
TSCA TSCA

RUO – Research Use Only