missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals o-Cresolphthalein complexone, indicator grade

Catalog No. p-7043979
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0385903 1 g
AAL0385906 5 g
AAL0385914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0385903 Supplier Thermo Scientific Chemicals Supplier No. L0385903
Only null left

CAS: 2411-89-4 | C32H32N2O12 | 636.61 g/mol

Indicator for complexometric titration of Mg, Ca, Sr and Ba. It is used as an alkaline earth metal ion indicator. Suggested to be colorless at pH 11, but turns pink in the presence of alkaline earth metals. Also utilized in an auto analyzer to determine serum calcium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indicator for complexometric titration of Mg, Ca, Sr and Ba. It is used as an alkaline earth metal ion indicator. Suggested to be colorless at pH 11, but turns pink in the presence of alkaline earth metals. Also utilized in an auto analyzer to determine serum calcium.

Solubility
Soluble in NaOH (50 mg/ml), aqueous ammonia, and organic solvents. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 2411-89-4
Molecular Formula C32H32N2O12
Molecular Weight (g/mol) 636.61
MDL Number MFCD00005911
InChI Key IYZPEGVSBUNMBE-UHFFFAOYSA-N
Synonym o-cresolphthalein complexone, phthalein purple, cresolphthalexon, o-cresolphthalexon, phthalein complexon, o-cresolphthalein complexon, cresolphthalein complexon, metal phthalein, cresolphthalein complexone, unii-a4p6737i7f
PubChem CID 75485
IUPAC Name 2-[[5-[1-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxymethyl)amino]acetic acid
SMILES CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1

Specifications

Melting Point ∽198°C (decomposition)
Quantity 1 g
Beilstein 381994
Solubility Information Soluble in NaOH (50mg/ml),aqueous ammonia,and organic solvents. Insoluble in water.
Formula Weight 636.62
Grade Indicator
Chemical Name or Material o-Cresolphthalein complexone
EINECSNumber 219-318-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only