missing translation for 'onlineSavingsMsg'
Learn More

o-Cresol 99.0+%, TCI America™

Catalog No. C040225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C040225G 25 g
C0402500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C040225G Supplier TCI America Supplier No. C040225G

Chemical Identifiers

CAS 95-48-7
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002226
InChI Key QWVGKYWNOKOFNN-UHFFFAOYSA-N
Synonym o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
PubChem CID 335
ChEBI CHEBI:28054
IUPAC Name 2-methylphenol
SMILES CC1=CC=CC=C1O

Specifications

Color White-Yellow
Boiling Point 192°C
Quantity 25 g
UN Number 3455
Formula Weight 108.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material o-Cresol
EINECSNumber (3)-0499&(4)-0057
RTECSNumber GO6300000
TSCA Yes