missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Cresol, MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 36922250MG
Description
Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal CareSpecifications
| 1110°F | |
| 0.3 mmHg (20°C) | |
| 81.0°C - Closed Cup | |
| CH3C6H4 OH | |
| 29°C to 31°C | |
| 2-Methylphenol | |
| 250 mg | |
| Analytical Standard |
| 95-48-7 | |
| 3.72 (vs. air) | |
| Neat | |
| MFCD00002226 | |
| 191°C | |
| Analytical Standard | |
| 108.14 |