Learn More
o-Cresol, 99%
CAS: 95-48-7 | C7H8O | 108.14 g/mol
Supplier: Thermo Scientific Chemicals 110550025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| o-Cresol | |
| 98.5 | |
| 30.0°C to 32.0°C | |
| 1.0400g/mL | |
| 81°C | |
| 98.5% min. (GC) | |
| C7H8O | |
| MFCD00002226 | |
| 06,349 | |
| 15,2564 | |
| Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform | |
| CC1=CC=CC=C1O | |
| 108.14 | |
| 4.75 mPa.s (35°C) | |
| 108.14 | |
| Liquid or Low Melting Solid |
| 95-48-7,108-95-2 | |
| 100.0 | |
| Amber to Colorless | |
| 191.0°C | |
| Authentic | |
| Glass bottle | |
| CH3C6H4OH | |
| 2.5 kg | |
| 1.04 | |
| o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene | |
| QWVGKYWNOKOFNN-UHFFFAOYSA-N | |
| 2-methylphenol | |
| 335 | |
| CHEBI:28054 | |
| 99% |
Chemical Identifiers
| 95-48-7 | |
| 108.14 | |
| QWVGKYWNOKOFNN-UHFFFAOYSA-N | |
| 335 | |
| 2-methylphenol |
| C7H8O | |
| MFCD00002226 | |
| o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene | |
| CHEBI:28054 | |
| CC1=CC=CC=C1O |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 202-423-8
Recommended Storage : Darkens on storage and exposure to light and air
RUO – Research Use Only