missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Chloranil 97.0+%, TCI America™
$463.97 - $1761.52
Chemical Identifiers
| CAS | 2435-53-2 |
|---|---|
| Molecular Formula | C6Cl4O2 |
| Molecular Weight (g/mol) | 245.86 |
| MDL Number | MFCD00001646 |
| InChI Key | VRGCYEIGVVTZCC-UHFFFAOYSA-N |
| Synonym | o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil |
| PubChem CID | 73252 |
| IUPAC Name | tetrachlorocyclohexa-3,5-diene-1,2-dione |
| SMILES | ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl |
Chemical Identifiers
| 2435-53-2 | |
| 245.86 | |
| VRGCYEIGVVTZCC-UHFFFAOYSA-N | |
| 73252 | |
| ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl |
| C6Cl4O2 | |
| MFCD00001646 | |
| o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil | |
| tetrachlorocyclohexa-3,5-diene-1,2-dione |
Specifications
| 2435-53-2 | |
| C6Cl4O2 | |
| 5 g | |
| VRGCYEIGVVTZCC-UHFFFAOYSA-N | |
| tetrachlorocyclohexa-3,5-diene-1,2-dione | |
| 73252 | |
| ≥97.0% (HPLC,T) | |
| o-Chloranil |
| 128°C | |
| MFCD00001646 | |
| o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil | |
| ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl | |
| 245.86 | |
| 245.86 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes