missing translation for 'onlineSavingsMsg'
Learn More

O-Benzylhydroxylamine Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America B10795G

 View more versions of this product

Catalog No. B10795G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

O-Benzylhydroxylamine Hydrochloride
White
MFCD00012952
o-benzylhydroxylamine hydrochloride, benzyloxyamine hydrochloride, o-benzylhydroxylamine hcl, o-benzyl-hydroxylamine hydrochloride, hydroxylamine, o-phenylmethyl-, hydrochloride, hydroxylamine, o-benzyl-, hydrochloride, o-benzyl-hydroxylamine hcl, o-benzyl hydroxylamine hydrochloride, aminooxy methyl benzene hydrochloride, oxybenzylamine, chloride
C1=CC=C(C=C1)CON.Cl
159.613
159.61
Crystalline Powder
2687-43-6
C7H10ClNO
5 g
HYDZPXNVHXJHBG-UHFFFAOYSA-N
O-benzylhydroxylamine;hydrochloride
102312
≥98.0% (T)

Chemical Identifiers

2687-43-6
159.613
HYDZPXNVHXJHBG-UHFFFAOYSA-N
102312
C1=CC=C(C=C1)CON.Cl
C7H10ClNO
MFCD00012952
o-benzylhydroxylamine hydrochloride, benzyloxyamine hydrochloride, o-benzylhydroxylamine hcl, o-benzyl-hydroxylamine hydrochloride, hydroxylamine, o-phenylmethyl-, hydrochloride, hydroxylamine, o-benzyl-, hydrochloride, o-benzyl-hydroxylamine hcl, o-benzyl hydroxylamine hydrochloride, aminooxy methyl benzene hydrochloride, oxybenzylamine, chloride
O-benzylhydroxylamine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : NC3040000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions