missing translation for 'onlineSavingsMsg'
Learn More

O-Benzylhydroxylamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7118838
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10795G 5 g
B107925G 25 g
B1079250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B10795G Supplier TCI America Supplier No. B10795G
Only null left

Chemical Identifiers

CAS 2687-43-6
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.613
MDL Number MFCD00012952
InChI Key HYDZPXNVHXJHBG-UHFFFAOYSA-N
Synonym o-benzylhydroxylamine hydrochloride, benzyloxyamine hydrochloride, o-benzylhydroxylamine hcl, o-benzyl-hydroxylamine hydrochloride, hydroxylamine, o-phenylmethyl-, hydrochloride, hydroxylamine, o-benzyl-, hydrochloride, o-benzyl-hydroxylamine hcl, o-benzyl hydroxylamine hydrochloride, aminooxy methyl benzene hydrochloride, oxybenzylamine, chloride
PubChem CID 102312
IUPAC Name O-benzylhydroxylamine;hydrochloride
SMILES C1=CC=C(C=C1)CON.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 159.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Benzylhydroxylamine Hydrochloride
RTECSNumber NC3040000
TSCA No