Learn More
O-Benzylhydroxylamine hydrochloride, 99%
CAS: 2687-43-6 | C7H10ClNO | 159.613 g/mol
Supplier: Thermo Scientific Chemicals A1739314
Description
For use in the formation of nitrones from carbonyl compounds and their use in cycloaddition reactions. Used as a reagent used in the synthesis of hydroxylamines and hydroxyamates. Effective reagent used to prepare α-hydroxybenzylamines from α-hydroxyketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFor use in the formation of nitrones from carbonyl compounds and their use in cycloaddition reactions. Used as a reagent used in the synthesis of hydroxylamines and hydroxyamates. Effective reagent used to prepare α-hydroxybenzylamines from α-hydroxyketones.
Solubility
Soluble in DMSO and methanol.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
Specifications
| O-Benzylhydroxylamine hydrochloride | |
| ∼238°C (sublimation) | |
| MFCD00012952 | |
| 3595440 | |
| o-benzylhydroxylamine hydrochloride, benzyloxyamine hydrochloride, o-benzylhydroxylamine hcl, o-benzyl-hydroxylamine hydrochloride, hydroxylamine, o-phenylmethyl-, hydrochloride, hydroxylamine, o-benzyl-, hydrochloride, o-benzyl-hydroxylamine hcl, o-benzyl hydroxylamine hydrochloride, aminooxy methyl benzene hydrochloride, oxybenzylamine, chloride | |
| HYDZPXNVHXJHBG-UHFFFAOYSA-N | |
| O-benzylhydroxylamine;hydrochloride | |
| 102312 | |
| 99% |
| 2687-43-6 | |
| C7H10ClNO | |
| 25 g | |
| Hygroscopic | |
| Soluble in DMSO and methanol. | |
| C1=CC=C(C=C1)CON.Cl | |
| 159.613 | |
| 159.62 |
Chemical Identifiers
| 2687-43-6 | |
| 159.613 | |
| HYDZPXNVHXJHBG-UHFFFAOYSA-N | |
| 102312 | |
| C1=CC=C(C=C1)CON.Cl |
| C7H10ClNO | |
| MFCD00012952 | |
| o-benzylhydroxylamine hydrochloride, benzyloxyamine hydrochloride, o-benzylhydroxylamine hcl, o-benzyl-hydroxylamine hydrochloride, hydroxylamine, o-phenylmethyl-, hydrochloride, hydroxylamine, o-benzyl-, hydrochloride, o-benzyl-hydroxylamine hcl, o-benzyl hydroxylamine hydrochloride, aminooxy methyl benzene hydrochloride, oxybenzylamine, chloride | |
| O-benzylhydroxylamine;hydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-249-0
RTECSNumber : NC3040000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only