missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine 98.0+%, TCI America™
$123.03 - $395.17
Chemical Identifiers
| CAS | 47173-80-8 |
|---|---|
| Molecular Formula | C15H20NO5 |
| Molecular Weight (g/mol) | 294.33 |
| MDL Number | MFCD00038248 |
| InChI Key | DMBKPDOAQVGTST-GFCCVEGCSA-M |
| Synonym | n-boc-o-benzyl-d-serine, boc-d-ser bzl-oh, boc-o-benzyl-d-serine, o-benzyl-n-tert-butoxycarbonyl-d-serine, d-serine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2r-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid, pubchem12444, pubchem14944, boc-d-ser obn-oh |
| PubChem CID | 2733693 |
| IUPAC Name | (2R)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}propanoate |
| SMILES | CC(C)(C)OC(=O)N[C@H](COCC1=CC=CC=C1)C([O-])=O |
Chemical Identifiers
| 47173-80-8 | |
| 294.33 | |
| DMBKPDOAQVGTST-GFCCVEGCSA-M | |
| 2733693 | |
| CC(C)(C)OC(=O)N[C@H](COCC1=CC=CC=C1)C([O-])=O |
| C15H20NO5 | |
| MFCD00038248 | |
| n-boc-o-benzyl-d-serine, boc-d-ser bzl-oh, boc-o-benzyl-d-serine, o-benzyl-n-tert-butoxycarbonyl-d-serine, d-serine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2r-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid, pubchem12444, pubchem14944, boc-d-ser obn-oh | |
| (2R)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}propanoate |
Specifications
| 47173-80-8 | |
| White | |
| MFCD00038248 | |
| n-boc-o-benzyl-d-serine, boc-d-ser bzl-oh, boc-o-benzyl-d-serine, o-benzyl-n-tert-butoxycarbonyl-d-serine, d-serine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2r-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid, pubchem12444, pubchem14944, boc-d-ser obn-oh | |
| CC(C)(C)OC(=O)N[C@H](COCC1=CC=CC=C1)C([O-])=O | |
| 294.33 | |
| 295.34 | |
| Crystalline Powder |
| 60°C | |
| C15H20NO5 | |
| 1 g | |
| DMBKPDOAQVGTST-GFCCVEGCSA-M | |
| (2R)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}propanoate | |
| 2733693 | |
| ≥98.0% (HPLC,T) | |
| O-Benzyl-N-(tert-butoxycarbonyl)-D-serine |
Safety and Handling
TSCA : No