missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl-N-(tert-butoxycarbonyl)-D-serine 98.0+%, TCI America™

Catalog No. B18635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18635G 5 g
B18631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18635G Supplier TCI America Supplier No. B18635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 47173-80-8
Molecular Formula C15H20NO5
Molecular Weight (g/mol) 294.33
MDL Number MFCD00038248
InChI Key DMBKPDOAQVGTST-GFCCVEGCSA-M
Synonym n-boc-o-benzyl-d-serine, boc-d-ser bzl-oh, boc-o-benzyl-d-serine, o-benzyl-n-tert-butoxycarbonyl-d-serine, d-serine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2r-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid, pubchem12444, pubchem14944, boc-d-ser obn-oh
PubChem CID 2733693
IUPAC Name (2R)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}propanoate
SMILES CC(C)(C)OC(=O)N[C@H](COCC1=CC=CC=C1)C([O-])=O

Specifications

Melting Point 60°C
Color White
Quantity 5 g
Formula Weight 295.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material O-Benzyl-N-(tert-butoxycarbonyl)-D-serine