missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl-N,N'-diisopropylisourea 97.0+%, TCI America™

Catalog No. B44801ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44801ML 1 mL
B44805ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44801ML Supplier TCI America Supplier No. B44801ML

Chemical Identifiers

CAS 2978-10-1
Molecular Formula C14H22N2O
Molecular Weight (g/mol) 234.34
InChI Key HTJDVAQGUYGUON-UHFFFAOYSA-N
PubChem CID 11128254
IUPAC Name benzyl N,N'-di(propan-2-yl)carbamimidate
SMILES CC(C)NC(=NC(C)C)OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 105°C
Quantity 1 mL
Synonym Benzyl N,N′C-Diisopropylcarbamimidate, N,N′C-Diisopropylcarbamimidic Acid Benzyl Ester
Formula Weight 234.34
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material O-Benzyl-N,N′-diisopropylisourea