accessibility menu, dialog, popup

UserName

CH3CH3HNNH3CCH3O(E/Z)

CAS RN 2978-10-1

IUPAC Name: N,N'-bis(propan-2-yl)(benzyloxy)methanimidamide
Synonyms: N,N'-diisopropylbenzyloxymethanimidamide
Molecular Weight (g/mol): 234.34
Molecular Formula: C14H22N2O
InChi Key: HTJDVAQGUYGUON-UHFFFAOYSA-N
SMILES: CC(C)NC(OCC1=CC=CC=C1)=NC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results