missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl-L-tyrosine, 98%

CAS: 16652-64-5 | C16H17NO3 | 271.32 g/mol

$118.03 - $367.34

Chemical Identifiers

CAS 16652-64-5
Molecular Formula C16H17NO3
Molecular Weight (g/mol) 271.32
MDL Number MFCD00002605
InChI Key KAFHLONDOVSENM-GGYSOQFKNA-N
Synonym o-benzyl-l-tyrosine, h-tyr bzl-oh, 4-benzyloxy-l-phenylalanine, s-2-amino-3-4-benzyloxy phenyl propanoic acid, o-benzyl-l-tyr, tyrosine, o-phenylmethyl, l-tyrosine benzyl ether, 2s-2-amino-3-4-benzyloxy phenyl propanoic acid, h-tyr bn-oh
PubChem CID 725701
IUPAC Name (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6367506
View Documents
Thermo Scientific Chemicals
H6367506
5 g
Each for $118.03
Only null left
 
AAH6367514
View Documents
Thermo Scientific Chemicals
H6367514
25 g
Each for $367.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16652-64-5
271.32
KAFHLONDOVSENM-GGYSOQFKNA-N
725701
N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
C16H17NO3
MFCD00002605
o-benzyl-l-tyrosine, h-tyr bzl-oh, 4-benzyloxy-l-phenylalanine, s-2-amino-3-4-benzyloxy phenyl propanoic acid, o-benzyl-l-tyr, tyrosine, o-phenylmethyl, l-tyrosine benzyl ether, 2s-2-amino-3-4-benzyloxy phenyl propanoic acid, h-tyr bn-oh
(2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoic acid

Specifications

259°C (decomposition)
White
MFCD00002605
o-benzyl-l-tyrosine, h-tyr bzl-oh, 4-benzyloxy-l-phenylalanine, s-2-amino-3-4-benzyloxy phenyl propanoic acid, o-benzyl-l-tyr, tyrosine, o-phenylmethyl, l-tyrosine benzyl ether, 2s-2-amino-3-4-benzyloxy phenyl propanoic acid, h-tyr bn-oh
N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
5 g
725701
98%
O-Benzyl-L-tyrosine
16652-64-5
C16H17NO3
2659642
KAFHLONDOVSENM-GGYSOQFKNA-N
(2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoic acid
271.32
271.31
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 240-699-1

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only