missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl-L-tyrosine, 98%

CAS: 16652-64-5 | C16H17NO3 | 271.32 g/mol

Supplier:  Thermo Scientific Chemicals H6367506

 View more versions of this product

Catalog No. AAH6367506

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

259°C (decomposition)
16652-64-5
C16H17NO3
2659642
KAFHLONDOVSENM-GGYSOQFKNA-N
(2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoic acid
271.32
271.31
Powder
O-Benzyl-L-tyrosine
White
MFCD00002605
o-benzyl-l-tyrosine, h-tyr bzl-oh, 4-benzyloxy-l-phenylalanine, s-2-amino-3-4-benzyloxy phenyl propanoic acid, o-benzyl-l-tyr, tyrosine, o-phenylmethyl, l-tyrosine benzyl ether, 2s-2-amino-3-4-benzyloxy phenyl propanoic acid, h-tyr bn-oh
N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
5 g
725701
98%

Chemical Identifiers

16652-64-5
271.32
KAFHLONDOVSENM-GGYSOQFKNA-N
725701
N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
C16H17NO3
MFCD00002605
o-benzyl-l-tyrosine, h-tyr bzl-oh, 4-benzyloxy-l-phenylalanine, s-2-amino-3-4-benzyloxy phenyl propanoic acid, o-benzyl-l-tyr, tyrosine, o-phenylmethyl, l-tyrosine benzyl ether, 2s-2-amino-3-4-benzyloxy phenyl propanoic acid, h-tyr bn-oh
(2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 240-699-1

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only