missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl-L-serine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. B14505G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14505G 5 g
B14501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14505G Supplier TCI America Supplier No. B14505G
Only null left

Chemical Identifiers

CAS 19525-87-2
Molecular Formula C11H16ClNO3
Molecular Weight (g/mol) 245.70
MDL Number MFCD00191312
InChI Key UDHSAWHYRXHQJP-UHFFFAOYNA-N
Synonym Methyl O-Benzyl-L-serinate Hydrochloride, H-Ser(Bzl)-OMe.HCl
PubChem CID 44630138
IUPAC Name methyl 2-amino-3-(benzyloxy)propanoate hydrochloride
SMILES Cl.COC(=O)C(N)COCC1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Formula Weight 245.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Benzyl-L-serine Methyl Ester Hydrochloride