accessibility menu, dialog, popup

UserName

CAS RN 19525-87-2

H3COOONH2HCl

CAS RN 19525-87-2

IUPAC Name: methyl 2-amino-3-(benzyloxy)propanoate hydrochloride
Synonyms: methyl 2-amino-3-(benzyloxy)propanoate hydrochloride
Molecular Weight (g/mol): 245.7
Molecular Formula: C11H16ClNO3
InChi Key: UDHSAWHYRXHQJP-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)COCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results