missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Benzyl-L-serine, 99%, Thermo Scientific™
Chemical Identifiers
| CAS | 4726-96-9 |
|---|---|
| Molecular Formula | C10H13NO3 |
| Molecular Weight (g/mol) | 195.22 |
| InChI Key | IDGQXGPQOGUGIX-VIFPVBQESA-N |
| Synonym | o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine |
| PubChem CID | 78457 |
| IUPAC Name | (2S)-2-amino-3-phenylmethoxypropanoic acid |
| SMILES | C1=CC=C(C=C1)COCC(C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC440930050
|
Thermo Scientific Chemicals
440930050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC440930250
|
Thermo Scientific Chemicals
440930250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 4726-96-9 | |
| 195.22 | |
| o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine | |
| (2S)-2-amino-3-phenylmethoxypropanoic acid |
| C10H13NO3 | |
| IDGQXGPQOGUGIX-VIFPVBQESA-N | |
| 78457 | |
| C1=CC=C(C=C1)COCC(C(=O)O)N |
Specifications
| 4726-96-9 | |
| 100.0 | |
| 98.5% min. (HPLC) | |
| C10H13NO3 | |
| 22.5 | |
| IDGQXGPQOGUGIX-VIFPVBQESA-N | |
| (2S)-2-amino-3-phenylmethoxypropanoic acid | |
| 78457 | |
| 99% |
| 98.5 | |
| Authentic | |
| Glass bottle | |
| 5g | |
| o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine | |
| C1=CC=C(C=C1)COCC(C(=O)O)N | |
| 195.22 | |
| 195.22 | |
| O-Benzyl-L-serine |
Safety and Handling
EINECSNumber : 225-220-6
RUO â Research Use Only