missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl-DL-serine 98.0+%, TCI America™

Catalog No. B08601G
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08601G 1 g
B086010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08601G Supplier TCI America Supplier No. B08601G

Chemical Identifiers

CAS 5445-44-3
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00021724,MFCD00065936
InChI Key IDGQXGPQOGUGIX-UHFFFAOYNA-N
Synonym o-benzyl-dl-serine, 2-amino-3-benzyloxy propanoic acid, o-benzylserine, h-dl-ser bzl-oh, 3-benzyloxy-dl-alanine, benzylserine, 2-amino-3-phenylmethoxy propanoic acid, o-benzyl-serine, acmc-1artk, spectrum2_001351
PubChem CID 4573
IUPAC Name 2-amino-3-(benzyloxy)propanoic acid
SMILES NC(COCC1=CC=CC=C1)C(O)=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 195.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Benzyl-DL-serine