accessibility menu, dialog, popup

UserName

NH2OOHO

CAS RN 5445-44-3

IUPAC Name: 2-amino-3-(benzyloxy)propanoic acid
Synonyms: benzylserine
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: IDGQXGPQOGUGIX-UHFFFAOYNA-N
SMILES: NC(COCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results