missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Benzoyl-N-tert-butylhydroxylamine Hydrochloride 98.0+%, TCI America™
$82.79 - $267.61
Chemical Identifiers
| CAS | 66809-86-7 |
|---|---|
| Molecular Formula | C11H16ClNO2 |
| Molecular Weight (g/mol) | 229.704 |
| InChI Key | UAYHNCIDRZTSPO-UHFFFAOYSA-N |
| PubChem CID | 12439863 |
| IUPAC Name | (tert-butylamino) benzoate;hydrochloride |
| SMILES | CC(C)(C)NOC(=O)C1=CC=CC=C1.Cl |
Chemical Identifiers
| 66809-86-7 | |
| 229.704 | |
| 12439863 | |
| CC(C)(C)NOC(=O)C1=CC=CC=C1.Cl |
| C11H16ClNO2 | |
| UAYHNCIDRZTSPO-UHFFFAOYSA-N | |
| (tert-butylamino) benzoate;hydrochloride |
Specifications
| 66809-86-7 | |
| White-Yellow | |
| 1 g | |
| CC(C)(C)NOC(=O)C1=CC=CC=C1.Cl | |
| 229.704 | |
| 229.70 | |
| Crystalline Powder |
| 187°C | |
| C11H16ClNO2 | |
| UAYHNCIDRZTSPO-UHFFFAOYSA-N | |
| (tert-butylamino) benzoate;hydrochloride | |
| 12439863 | |
| ≥98.0% (HPLC,N) | |
| O-Benzoyl-N-tert-butylhydroxylamine Hydrochloride |
Safety and Handling
TSCA : No