accessibility menu, dialog, popup

UserName

H3CH3CCH3NHOOHCl

CAS RN 66809-86-7

IUPAC Name: tert-butylamino benzoate hydrochloride
Synonyms: tert-butylamino benzoate hydrochloride
Molecular Weight (g/mol): 229.7
Molecular Formula: C11H16ClNO2
InChi Key: UAYHNCIDRZTSPO-UHFFFAOYSA-N
SMILES: Cl.CC(C)(C)NOC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)