missing translation for 'onlineSavingsMsg'
Learn More

O-Benzoyl-N-tert-butylhydroxylamine Hydrochloride 98.0+%, TCI America™

Catalog No. B32381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32381G 1 g
B32385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32381G Supplier TCI America Supplier No. B32381G

Chemical Identifiers

CAS 66809-86-7
Molecular Formula C11H16ClNO2
Molecular Weight (g/mol) 229.704
InChI Key UAYHNCIDRZTSPO-UHFFFAOYSA-N
PubChem CID 12439863
IUPAC Name (tert-butylamino) benzoate;hydrochloride
SMILES CC(C)(C)NOC(=O)C1=CC=CC=C1.Cl

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 1 g
Formula Weight 229.70
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material O-Benzoyl-N-tert-butylhydroxylamine Hydrochloride