missing translation for 'onlineSavingsMsg'
Learn More

o-Anisidine 98.0+%, TCI America™

Catalog No. p-7116928
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A048625ML 25 mL
A0486500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A048625ML Supplier TCI America Supplier No. A048625ML
Only null left

Chemical Identifiers

CAS 90-04-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007688
InChI Key VMPITZXILSNTON-UHFFFAOYSA-N
Synonym o-anisidine, 2-anisidine, ortho-anisidine, 2-aminoanisole, o-methoxyaniline, o-aminoanisole, benzenamine, 2-methoxy, 2-methoxybenzenamine, o-methoxyphenylamine, o-anisylamine
PubChem CID 7000
ChEBI CHEBI:82288
IUPAC Name 2-methoxyaniline
SMILES COC1=CC=CC=C1N

Specifications

Color Yellow
Boiling Point 225°C
Quantity 25 mL
UN Number 2431
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material o-Anisidine
EINECSNumber (3)-0682
RTECSNumber BZ5410000
TSCA Yes
Recommended Storage Refrigerator