missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Anisidine, 99%
Description
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.
Solubility
Soluble in dilute inorganic acid, alcohol, diethyl ether. Not miscible in water.
Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents, acid anhydrides, chloroformates, acids.
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.
Solubility
Soluble in dilute inorganic acid, alcohol, diethyl ether. Not miscible in water.
Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents, acid anhydrides, chloroformates, acids.
Specifications
Specifications
| Melting Point | 5°C to 6°C |
| Density | 1.092 g/mL |
| Boiling Point | 224°C to 225°C |
| Flash Point | 98°C (208°F) |
| Assay Percent Range | 99% |
| Refractive Index | 1.574 |
| Quantity | 250 g |
| UN Number | UN2431 |
| Beilstein | 386210 |
| Sensitivity | Light sensitive |
| Show More |
Safety and Handling
| Hazard Category | H301+H311-H341-H350-H373 |
| Hazard Statement | GHS H Statement H301-H311-H331-H350-H341 |
| Precautionary Statement | P201-P202-P260-P264b-P270-P281-P301+P310-P302+P352-P308+P313-P312-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANISIDINES |
| EINECSNumber | 201-963-1 |
| RTECSNumber | BZ5410000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only