missing translation for 'onlineSavingsMsg'
Learn More

o-Anisic Acid 98.0+%, TCI America™

Catalog No. p-7116924
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A048125G 25 g
A0481500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A048125G Supplier TCI America Supplier No. A048125G

Chemical Identifiers

CAS 579-75-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002431
InChI Key ILUJQPXNXACGAN-UHFFFAOYSA-N
Synonym o-anisic acid, o-methoxybenzoic acid, 2-anisic acid, o-methylsalicylic acid, benzoic acid, 2-methoxy, salicylic acid methyl ether, 2-methoxy-benzoic acid, 2-methoxybenzoicacid, o-methoxy benzoic acid, ortho-methoxybenzoic acid
PubChem CID 11370
ChEBI CHEBI:421840
IUPAC Name 2-methoxybenzoic acid
SMILES COC1=CC=CC=C1C(=O)O

Specifications

Melting Point 103°C
Color White
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material o-Anisic Acid
EINECSNumber (3)-1659
RTECSNumber BZ4385000
TSCA Yes