missing translation for 'onlineSavingsMsg'
Learn More

o-Anisaldehyde 98.0+%, TCI America™

Catalog No. A047925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A047925G 25 g
A0479100G 100 g
A0479500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A047925G Supplier TCI America Supplier No. A047925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-02-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00003308
InChI Key PKZJLOCLABXVMC-UHFFFAOYSA-N
Synonym o-anisaldehyde, benzaldehyde, 2-methoxy, 2-anisaldehyde, o-methoxybenzaldehyde, 6-methoxybenzaldehyde, salicylaldehyde methyl ether, 2-methoxybenzenecarboxaldehyde, benzaldehyde, o-methoxy, 2-methoxyphenylformaldehyde, formylanisole, o
PubChem CID 8658
IUPAC Name 2-methoxybenzaldehyde
SMILES COC1=CC=CC=C1C=O

Specifications

Color White-Yellow
Boiling Point 243°C
Quantity 25 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material o-Anisaldehyde
EINECSNumber (3)-2661&(9)-1879
RTECSNumber BZ2610000
TSCA Yes
Recommended Storage Refrigerator