missing translation for 'onlineSavingsMsg'
Learn More

O-Acetylsalicyloyl Chloride 97.0+%, TCI America™

Catalog No. A19175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19175G 5 g
A191725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19175G Supplier TCI America Supplier No. A19175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5538-51-2
Molecular Formula C9H7ClO3
Molecular Weight (g/mol) 198.602
MDL Number MFCD00000663
InChI Key DSGKWFGEUBCEIE-UHFFFAOYSA-N
Synonym 2-acetoxybenzoyl chloride, o-acetylsalicyloyl chloride, acetylsalicyloyl chloride, 2-chlorocarbonyl phenyl acetate, benzoyl chloride, 2-acetyloxy, aspirin chloride, o-acetylsalicyloylchloride, acetylsalicoyl chloride, 2-carbonochloridoyl phenyl acetate, ascc
PubChem CID 79668
IUPAC Name (2-carbonochloridoylphenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1C(=O)Cl

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 135°C
Quantity 5 g
UN Number 3261
Formula Weight 198.60
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material O-Acetylsalicyloyl Chloride