missing translation for 'onlineSavingsMsg'
Learn More

O-Acetylsalicyloyl chloride, 97%

Catalog No. p-7043472
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0091914 25 g
AAL0091922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0091914 Supplier Thermo Scientific Chemicals Supplier No. L0091914

CAS: 5538-51-2 | C9H7ClO3 | 198.602 g/mol

O-Acetylsalicyloyl chloride was used in the synthesis of enantiomerically pure bidentate heteroorganic ligands built on simple achiral skeletons and containing an aziridine moiety. It was used in the general synthesis of acetoxybenzamides. It was used in the chemical synthesis of 2,2,6,6-tetramethyl-1-piperidinyloxy(TEMPO)-aspirin conjugate via condensation reaction with 4-hydroxy-TEMPO. It was used as reagent in acylation of cyclobutenediones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Acetylsalicyloyl chloride was used in the synthesis of enantiomerically pure bidentate heteroorganic ligands built on simple achiral skeletons and containing an aziridine moiety. It was used in the general synthesis of acetoxybenzamides. It was used in the chemical synthesis of 2,2,6,6-tetramethyl-1-piperidinyloxy(TEMPO)-aspirin conjugate via condensation reaction with 4-hydroxy-TEMPO. It was used as reagent in acylation of cyclobutenediones.

Solubility
Soluble in toluene. Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from strong bases.

Chemical Identifiers

CAS 5538-51-2
Molecular Formula C9H7ClO3
Molecular Weight (g/mol) 198.602
MDL Number MFCD00000663
InChI Key DSGKWFGEUBCEIE-UHFFFAOYSA-N
Synonym 2-acetoxybenzoyl chloride, o-acetylsalicyloyl chloride, acetylsalicyloyl chloride, 2-chlorocarbonyl phenyl acetate, benzoyl chloride, 2-acetyloxy, aspirin chloride, o-acetylsalicyloylchloride, acetylsalicoyl chloride, 2-carbonochloridoyl phenyl acetate, ascc
PubChem CID 79668
IUPAC Name (2-carbonochloridoylphenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1C(=O)Cl

Specifications

Melting Point 47°C to 50°C
Boiling Point 134°C to 136°C (12 mmHg)
Flash Point >110°C (230°F)
Odor Pungent, Makes Eyes Water
Quantity 25 g
UN Number UN3261
Beilstein 880372
Sensitivity Moisture sensitive
Solubility Information Soluble in toluene. Reacts with water.
Formula Weight 198.61
Percent Purity 97%
Chemical Name or Material O-Acetylsalicyloyl chloride
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 226-899-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only