missing translation for 'onlineSavingsMsg'
Learn More

O-Acetyl-N-tert-butylhydroxylamine Hydrochloride 98.0+%, TCI America™

Catalog No. A21381G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21381G Supplier TCI America Supplier No. A21381G
Only null left

Chemical Identifiers

CAS 851074-40-3
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.633
InChI Key CWIOQGVXNNMFJC-UHFFFAOYSA-N
PubChem CID 44629988
IUPAC Name (tert-butylamino) acetate;hydrochloride
SMILES CC(=O)ONC(C)(C)C.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 167.63
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material O-Acetyl-N-tert-butylhydroxylamine Hydrochloride
TSCA No
Recommended Storage Refrigerator