accessibility menu, dialog, popup

UserName

CAS RN 851074-40-3

H3COONHCH3CH3H3CHCl

CAS RN 851074-40-3

IUPAC Name: tert-butylamino acetate hydrochloride
Synonyms: tert-butylamino acetate hydrochloride
Molecular Weight (g/mol): 167.63
Molecular Formula: C6H14ClNO2
InChi Key: CWIOQGVXNNMFJC-UHFFFAOYSA-N
SMILES: Cl.CC(=O)ONC(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results