missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Acetyl-N-methylhydroxylamine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America A23651G
Specifications
| O-Acetyl-N-methylhydroxylamine Hydrochloride | |
| 111°C | |
| C3H8ClNO2 | |
| ZTMICKVBCKWFHL-UHFFFAOYSA-N | |
| methylamino acetate;hydrochloride | |
| 13675437 | |
| ≥98.0% (N,T) |
| 19689-95-3 | |
| White-Yellow | |
| 1 g | |
| CC(=O)ONC.Cl | |
| 125.552 | |
| 125.55 | |
| Crystalline Powder |
Chemical Identifiers
| 19689-95-3 | |
| 125.552 | |
| 13675437 | |
| CC(=O)ONC.Cl |
| C3H8ClNO2 | |
| ZTMICKVBCKWFHL-UHFFFAOYSA-N | |
| methylamino acetate;hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator