missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Acetyl-N-methylhydroxylamine Hydrochloride 98.0+%, TCI America™
$229.29 - $777.32
Chemical Identifiers
| CAS | 19689-95-3 |
|---|---|
| Molecular Formula | C3H8ClNO2 |
| Molecular Weight (g/mol) | 125.552 |
| InChI Key | ZTMICKVBCKWFHL-UHFFFAOYSA-N |
| PubChem CID | 13675437 |
| IUPAC Name | methylamino acetate;hydrochloride |
| SMILES | CC(=O)ONC.Cl |
Chemical Identifiers
| 19689-95-3 | |
| 125.552 | |
| 13675437 | |
| CC(=O)ONC.Cl |
| C3H8ClNO2 | |
| ZTMICKVBCKWFHL-UHFFFAOYSA-N | |
| methylamino acetate;hydrochloride |
Specifications
| 19689-95-3 | |
| White-Yellow | |
| 1 g | |
| CC(=O)ONC.Cl | |
| 125.552 | |
| 125.55 | |
| Crystalline Powder |
| 111°C | |
| C3H8ClNO2 | |
| ZTMICKVBCKWFHL-UHFFFAOYSA-N | |
| methylamino acetate;hydrochloride | |
| 13675437 | |
| ≥98.0% (N,T) | |
| O-Acetyl-N-methylhydroxylamine Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator