missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals O-Acetyl-L-serine hydrochloride

Catalog No. p-7043004
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6493303 1 g
AAJ6493306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6493303 Supplier Thermo Scientific Chemicals Supplier No. J6493303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Substrate for cysteine synthase and beta-substituted alanine synthase in higher plants | CAS: 66638-22-0 | C5H10ClNO4 | 183.588 g/mol

O-Acetyl-L-serine, a cysteine precursor, is used as a substrate for the identification, differentiation and characterization of O-acetyl-L-serine(thiol)lyase(s) (OASTL) involved in cysteine biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Acetyl-L-serine, a cysteine precursor, is used as a substrate for the identification, differentiation and characterization of O-acetyl-L-serine(thiol)lyase(s) (OASTL) involved in cysteine biosynthesis.

Solubility
Soluble in ethanol almost transparency.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.

Chemical Identifiers

CAS 66638-22-0
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00060169
InChI Key MGQOSZSPKMBSRW-WCCKRBBISA-N
Synonym o-acetyl-l-serine hydrochloride, s-3-acetoxy-2-aminopropanoic acid hydrochloride, o-acetyl-l-serine hcl, h-ser ac-oh.hcl, o-acetyl-l-serine-hydrogen chloride 1/1
PubChem CID 44630053
IUPAC Name (2S)-3-acetyloxy-2-aminopropanoic acid;hydrochloride
SMILES CC(=O)OCC(C(=O)O)N.Cl

Specifications

Color White
Solubility Information Soluble in ethanol almost transparency.
Quantity 1 g
Formula Weight 183.59
Physical Form Powder
Chemical Name or Material O-Acetyl-L-serine hydrochloride
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only