accessibility menu, dialog, popup

UserName

H3COONH2OHO

CAS RN 66638-22-0

IUPAC Name: 3-(acetyloxy)-2-aminopropanoic acid
Synonyms: O-acetylserine
Molecular Weight (g/mol): 147.13
Molecular Formula: C5H9NO4
InChi Key: VZXPDPZARILFQX-UHFFFAOYNA-N
SMILES: CC(=O)OCC(N)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)