missing translation for 'onlineSavingsMsg'
Learn More

O-Acetyl-L-serine Hydrochloride 98.0+%, TCI America™

Catalog No. A0834100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0834100MG 100 mg
A08341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0834100MG Supplier TCI America Supplier No. A0834100MG

Chemical Identifiers

CAS 66638-22-0
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00060169
InChI Key MGQOSZSPKMBSRW-WCCKRBBISA-N
Synonym o-acetyl-l-serine hydrochloride, s-3-acetoxy-2-aminopropanoic acid hydrochloride, o-acetyl-l-serine hcl, h-ser ac-oh.hcl, o-acetyl-l-serine-hydrogen chloride 1/1
PubChem CID 44630053
IUPAC Name (2S)-3-acetyloxy-2-aminopropanoic acid;hydrochloride
SMILES CC(=O)OCC(C(=O)O)N.Cl

Specifications

Melting Point 163°C
Color White
Quantity 100 mg
Formula Weight 183.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Acetyl-L-serine Hydrochloride
TSCA No
Recommended Storage Refrigerator