missing translation for 'onlineSavingsMsg'
Learn More

O-(4-Methoxybenzyl)hydroxylamine hydrochloride, 98%

Catalog No. AAH6331203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6331203 1 g
AAH6331206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6331203 Supplier Thermo Scientific Chemicals Supplier No. H6331203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 876-33-5 | C8H12ClNO2 | 189.64 g/mol

O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 876-33-5
MDL Number MFCD01114582
Synonym o-4-methoxybenzyl hydroxylamine, o-4-methoxyphenyl methyl hydroxylamine, p-methoxybenzyloxyamine, hydroxylamine, o-p-methoxybenzyl, hydroxylamine, o-4-methoxyphenyl methyl, o-p-methoxybenzyl hydroxylamine, o-p-methoxybenzyl-hydroxylamine, o-4-methoxy-benzyl-hydroxylamine, 1-aminooxy methyl-4-methoxybenzene, 1-aminooxy methyl 4-methoxy-benzene
PubChem CID 120426
IUPAC Name O-[(4-methoxyphenyl)methyl]hydroxylamine

Specifications

Color White
Molecular Formula C8H11NO2
Quantity 1 g
InChI Key MVSMBIBGGPSEHQ-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)CON
Molecular Weight (g/mol) 153.181
Formula Weight 153.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material O-(4-Methoxybenzyl)hydroxylamine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber NC3676000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only