missing translation for 'onlineSavingsMsg'
Learn More

O-(2-Methoxyisopropyl)hydroxylamine 95.0+%, TCI America™

Catalog No. M20855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20855G 5 g
M20851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20855G Supplier TCI America Supplier No. M20855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103491-33-4
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
InChI Key HAWYTJZHQIXQCP-UHFFFAOYSA-N
PubChem CID 13608208
IUPAC Name O-(2-methoxypropan-2-yl)hydroxylamine
SMILES CC(C)(OC)ON

Specifications

Color Colorless
Boiling Point 73°C
Quantity 5 g
UN Number 2734
Formula Weight 105.14
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material O-(2-Methoxyisopropyl)hydroxylamine
TSCA No
Recommended Storage Refrigerator