accessibility menu, dialog, popup

UserName

CAS RN 103491-33-4

IUPAC Name: O-(2-methoxypropan-2-yl)hydroxylamine
Synonyms: O-(2-methoxypropan-2-yl)hydroxylamine
Molecular Weight (g/mol): 105.14
Molecular Formula: C4H11NO2
InChi Key: HAWYTJZHQIXQCP-UHFFFAOYSA-N
SMILES: COC(C)(C)ON
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results